Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008503.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008503.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008503.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008503.hg.1 |
TC20000577.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000158110.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158110.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158110.hg.1 |
TC2000008231.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158110.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158110.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158108.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158108.hg.1 |
TC2000008231.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158108.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008231.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158111.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158111.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158111.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158111.hg.1 |
NM_015417.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158111.hg.1 |
TC2000008231.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158111.hg.1 |
ENST00000379756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158111.hg.1 |
ENST00000463490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158111.hg.1 |
ENST00000471499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158111.hg.1 |
NM_015417.4 |
Refseq |