Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016694.hg.1 |
TC0X001233.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016694.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016694.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170770.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170770.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170770.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170770.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170770.hg.1 |
TC0X00010399.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010399.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170771.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170771.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170771.hg.1 |
ENST00000372728.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170771.hg.1 |
NM_018476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170771.hg.1 |
TC0X00010399.hg.1 |
Clariom D TC |