Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022409.hg.1 |
TC08001703.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022409.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022409.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022408.hg.1 |
TC08001703.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022408.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022408.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012085.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012085.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012085.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012085.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012085.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012085.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012085.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012085.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012085.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012085.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012085.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012085.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012085.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012085.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012085.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012085.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012085.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012085.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012085.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012085.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182178.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182178.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182178.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182178.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182178.hg.1 |
TC0800012085.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800182177.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182177.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182177.hg.1 |
ENST00000246515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182177.hg.1 |
NM_020427.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182177.hg.1 |
TC0800012085.hg.1 |
Clariom D TC |