Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000491173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000491173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000491173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000491173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000491173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000491173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |