Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014687.hg.1 |
ENST00000439132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014687.hg.1 |
ENST00000439132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014687.hg.1 |
ENST00000439132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014687.hg.1 |
ENST00000439132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014687.hg.1 |
ENST00000439132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014687.hg.1 |
ENST00000439132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014687.hg.1 |
ENST00000439132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014687.hg.1 |
ENST00000439132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000347159.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
ENST00000376588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_021154.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007667.hg.1 |
NM_058179.3 |
Refseq |