Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008124.hg.1 |
ENST00000411890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008124.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008124.hg.1 |
TC17000652.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008128.hg.1 |
ENST00000240306.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008128.hg.1 |
ENST00000411890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008128.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008128.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008128.hg.1 |
TC17000652.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008125.hg.1 |
ENST00000411890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008125.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008125.hg.1 |
TC17000652.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000503410.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000503410.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000503410.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000503410.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000503410.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000505318.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000505318.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000505318.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000505318.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000505318.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008126.hg.1 |
ENST00000411890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008126.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008126.hg.1 |
TC17000652.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700160831.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160831.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160831.hg.1 |
ENST00000503410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160831.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160831.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160831.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160831.hg.1 |
ENST00000503410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160831.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160831.hg.1 |
TC1700008225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000503410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000503410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000503410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000503410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000503410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000505318.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000505318.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000505318.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000505318.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000505318.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008225.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160834.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160834.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160834.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160834.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160834.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160834.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160834.hg.1 |
TC1700008225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700160837.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160837.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160837.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160837.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160837.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160837.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160837.hg.1 |
ENST00000240306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160837.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160837.hg.1 |
ENST00000503276.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160837.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160837.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160837.hg.1 |
NM_138281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160837.hg.1 |
TC1700008225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700160833.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160833.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160833.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160833.hg.1 |
ENST00000411890.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160833.hg.1 |
ENST00000611342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160833.hg.1 |
NM_001934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160833.hg.1 |
TC1700008225.hg.1 |
Clariom D TC |