Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001800.hg.1 |
TC03000085.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001800.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001800.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013249.hg.1 |
ENST00000436601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008508.hg.1 |
ENST00000566307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011206.hg.1 |
ENST00000453255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008496.hg.1 |
ENST00000551246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001801.hg.1 |
TC03000085.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001801.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001801.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006669.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008867.hg.1 |
ENST00000502948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001799.hg.1 |
TC03000085.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001799.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001799.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001802.hg.1 |
TC03000085.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001802.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001802.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146482.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146482.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146482.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146482.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146482.hg.1 |
TC0300006669.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011206.hg.1 |
ENST00000453255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008867.hg.1 |
ENST00000502948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008496.hg.1 |
ENST00000551246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008508.hg.1 |
ENST00000566307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146478.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146478.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146478.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146478.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146478.hg.1 |
TC0300006669.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300146481.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146481.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146481.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146481.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146481.hg.1 |
TC0300006669.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200013249.hg.1 |
ENST00000436601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
ENST00000306024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
NM_014463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006669.hg.1 |
NM_014463.2 |
Refseq |