Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040288.1 | 7 | 1045 | 82 | ||
NR_040289.1 | 5 | 848 | 82 | |||
GenBank mRNA | AJ278012.1 | - | 6 | 831 | 82 | |
BC035351.1 | - | 5 | 715 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |