Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018110.hg.1 |
ENST00000357627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018110.hg.1 |
NM_001001966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061479.hg.1 |
TC01004079.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061479.hg.1 |
ENST00000357627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061479.hg.1 |
NM_001001966.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100236272.hg.1 |
ENST00000357627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100236272.hg.1 |
NM_001001966.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100236272.hg.1 |
ENST00000357627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100236272.hg.1 |
NM_001001966.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100236272.hg.1 |
TC0100018110.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018110.hg.1 |
ENST00000357627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018110.hg.1 |
NM_001001966.1 |
Refseq |