Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15009462.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15009462.hg.1 |
TC15001023.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15009460.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15009460.hg.1 |
TC15001023.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15009461.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15009461.hg.1 |
TC15001023.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15009459.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15009459.hg.1 |
TC15001023.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008661.hg.1 |
ENST00000629550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500162517.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500162517.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500162517.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500162517.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500162517.hg.1 |
TC1500008640.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008640.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008673.hg.1 |
ENST00000556855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500162520.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500162520.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500162520.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500162520.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500162520.hg.1 |
TC1500008640.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500162514.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500162514.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500162514.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500162514.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500162514.hg.1 |
TC1500008640.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500162516.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500162516.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500162516.hg.1 |
ENST00000604105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500162516.hg.1 |
NR_028506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500162516.hg.1 |
TC1500008640.hg.1 |
Clariom D TC |