Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR3178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036139.1 | 1 | 39 | 53 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cementum protein 1 | CEMP1 |
CP-23, CP23 |
-514.0 | Sense | Chr. 16 - 2530035 - 2531408 on NCBI Build GRCh38 |
Downstream | 3-phosphoinositide dependent protein kinase 1 | PDPK1 |
PDK1, PDPK2, PDPK2P, PRO0461 |
6.0kb | Anti-sense | Chr. 16 - 2537964 - 2603190 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |