Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000220856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000220856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000220856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000220856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000220856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000220856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000220856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000220856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000250160.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000250160.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000250160.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000250160.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000250160.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000250160.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000250160.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000250160.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000250160.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000250160.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000250160.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000250160.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000250160.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000250160.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000250160.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000377862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000377862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000377862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000377862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000377862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000377862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000377862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000377862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000377862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000517423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000517423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000517423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000517423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000517423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000517423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000519433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000519433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000519433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000519433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000519433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
ENST00000602893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204869.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204869.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204869.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204869.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204869.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204869.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204869.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204869.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204869.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204869.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204869.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204869.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204869.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204869.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_001204870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_003882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_080838.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_080838.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_080838.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_080838.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_080838.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_080838.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_080838.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_080838.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_080838.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_080838.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_080838.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_080838.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_080838.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_080838.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NM_080838.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NR_037944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NR_037944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NR_037944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NR_037944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NR_037944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NR_037944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NR_037944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NR_037944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NR_037944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NR_037944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NR_037944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NR_037944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008955.hg.1 |
NR_037944.1 |
Refseq |