Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000548872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000549124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000549124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000549124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000551146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000551146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000551146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000551146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000551146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
ENST00000551146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027499.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027499.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027499.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027499.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027499.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027499.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027499.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027499.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027499.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027499.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007582.hg.1 |
NR_027501.1 |
Refseq |