Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010358.hg.1 |
ENST00000544824.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010358.hg.1 |
ENST00000544824.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010358.hg.1 |
ENST00000544824.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010358.hg.1 |
ENST00000544824.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010358.hg.1 |
NM_001190489.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010358.hg.1 |
NM_001190489.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010358.hg.1 |
NM_001190489.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
NM_001190476.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
NM_001190476.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
NM_001190476.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009462.hg.1 |
ENST00000604952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009462.hg.1 |
ENST00000604952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009462.hg.1 |
ENST00000604952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009462.hg.1 |
ENST00000604952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009462.hg.1 |
ENST00000604952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014190.hg.1 |
TC07000914.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014190.hg.1 |
NM_001190487.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007183.hg.1 |
ENST00000582078.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007183.hg.1 |
ENST00000582078.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007183.hg.1 |
ENST00000607661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007183.hg.1 |
ENST00000607661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
NM_001190476.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
NM_001190476.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
NM_001190476.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010358.hg.1 |
ENST00000544824.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010358.hg.1 |
ENST00000544824.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010358.hg.1 |
ENST00000544824.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010358.hg.1 |
ENST00000544824.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010358.hg.1 |
NM_001190489.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010358.hg.1 |
NM_001190489.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010358.hg.1 |
NM_001190489.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007183.hg.1 |
ENST00000582078.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007183.hg.1 |
ENST00000582078.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007183.hg.1 |
ENST00000607661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007183.hg.1 |
ENST00000607661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009462.hg.1 |
ENST00000604952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009462.hg.1 |
ENST00000604952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009462.hg.1 |
ENST00000604952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009462.hg.1 |
ENST00000604952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009462.hg.1 |
ENST00000604952.1 |
Ensembl |