Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011482.hg.1 |
ENST00000227638.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011482.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011482.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011482.hg.1 |
TC11000916.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011484.hg.1 |
ENST00000227638.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011484.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011484.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011484.hg.1 |
TC11000916.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008797.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100164977.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100164977.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100164977.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100164977.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100164977.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100164977.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100164977.hg.1 |
TC1100008797.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100164976.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100164976.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100164976.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100164976.hg.1 |
ENST00000227638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100164976.hg.1 |
ENST00000436171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100164976.hg.1 |
NM_015368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100164976.hg.1 |
TC1100008797.hg.1 |
Clariom D TC |