Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
ENST00000374213.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
ENST00000374213.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
ENST00000374213.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
ENST00000374213.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
ENST00000470468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
ENST00000470468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
ENST00000470468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
ENST00000470468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
ENST00000492808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
ENST00000492808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
ENST00000492808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
ENST00000492808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
NM_001803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
NM_001803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
NM_001803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
NM_001803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007451.hg.1 |
NM_001803.2 |
Refseq |