Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040161.hg.1 |
TC02002532.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040161.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040161.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040159.hg.1 |
TC02002532.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040159.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040159.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040162.hg.1 |
TC02002532.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040162.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040162.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209166.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209166.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209166.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209166.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209166.hg.1 |
TC0200014938.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000419609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000422703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014938.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209167.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209167.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209167.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209167.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209167.hg.1 |
TC0200014938.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200209165.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209165.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209165.hg.1 |
ENST00000423106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209165.hg.1 |
NR_033882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209165.hg.1 |
TC0200014938.hg.1 |
Clariom D TC |