Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011233.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011233.hg.1 |
TC17000862.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009785.hg.1 |
ENST00000581753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000586217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000586217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000586217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000586217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000586217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000586217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000586217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000587258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000587258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000587258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000587258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000587258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000587501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000587501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000587501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000589790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000592074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000592074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000592074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000592629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000592629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000592629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000592629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000592629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
ENST00000592629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008923.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011234.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011234.hg.1 |
TC17000862.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700167177.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700167177.hg.1 |
ENST00000587501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700167177.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700167177.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700167177.hg.1 |
ENST00000587501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700167177.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700167177.hg.1 |
TC1700008923.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1900009785.hg.1 |
ENST00000581753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700167175.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700167175.hg.1 |
ENST00000587501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700167175.hg.1 |
ENST00000592074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700167175.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700167175.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700167175.hg.1 |
ENST00000587501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700167175.hg.1 |
ENST00000592074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700167175.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700167175.hg.1 |
TC1700008923.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000586217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000586217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000586217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000586217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000586217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000586217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000586217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000587258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000587258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000587258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000587258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000587258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000587501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000587501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000587501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000589790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000592074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000592074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000592074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000592629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000592629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000592629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000592629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000592629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
ENST00000592629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NM_001080419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008923.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700167178.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700167178.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700167178.hg.1 |
ENST00000586527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700167178.hg.1 |
NR_038131.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700167178.hg.1 |
TC1700008923.hg.1 |
Clariom D TC |