Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017589.3 | NP_060059.1 | 3 - 4 | 504 | 108 | |
XM_011542876.2 | XP_011541178.1 | 3 - 4 | 538 | 108 | ||
XM_011542878.2 | XP_011541180.1 | 3 - 4 | 538 | 108 | ||
XM_011542879.2 | XP_011541181.1 | 3 - 4 | 538 | 108 | ||
XM_011542880.2 | XP_011541182.1 | 3 - 4 | 538 | 108 | ||
GenBank mRNA | AJ271351.1 | - | 2 - 3 | 309 | 108 | |
BC031045.1 | - | 3 - 4 | 490 | 5271706 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |