Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKZF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257411.1 | NP_001244340.1 | 3 - 4 | 228 | 82 | |
GenBank mRNA | AY377977.1 | - | 3 - 4 | 228 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |