Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000611082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000611082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004890.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004890.hg.1 |
TC13000430.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004891.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004891.hg.1 |
TC13000430.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000611082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000611082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167262.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167262.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167262.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167262.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167262.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167262.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167262.hg.1 |
TC1300010008.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300167259.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167259.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167259.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167259.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167259.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167259.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167259.hg.1 |
TC1300010008.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300167260.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167260.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167260.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167260.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167260.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167260.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167260.hg.1 |
TC1300010008.hg.1 |
Clariom D TC |