Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009324.hg.1 |
ENST00000446391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009324.hg.1 |
ENST00000446391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009324.hg.1 |
ENST00000446391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009324.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009324.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009324.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009324.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009324.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162585.hg.1 |
ENST00000446391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162585.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162585.hg.1 |
ENST00000446391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162585.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162585.hg.1 |
TC1300009324.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300162588.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162588.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162588.hg.1 |
TC1300009324.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300162593.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162593.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162593.hg.1 |
TC1300009324.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300162583.hg.1 |
ENST00000446391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162583.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162583.hg.1 |
ENST00000446391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162583.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162583.hg.1 |
TC1300009324.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009324.hg.1 |
ENST00000446391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009324.hg.1 |
ENST00000446391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009324.hg.1 |
ENST00000446391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009324.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009324.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009324.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009324.hg.1 |
NR_046869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009324.hg.1 |
NR_046869.2 |
Refseq |