Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270837.1 | NP_001257766.1 | 7 - 8 | 1095 | 115 | |
NM_001270841.1 | NP_001257770.1 | 3 - 4 | 288 | 115 | ||
NM_001312.3 | NP_001303.1 | 7 - 8 | 651 | 115 | ||
NR_073081.1 | 5 - 6 | 452 | 115 | |||
NR_073082.1 | 7 - 8 | 627 | 115 | |||
NR_073083.1 | 6 - 7 | 556 | 115 | |||
NR_073084.1 | 7 - 8 | 1404 | 115 | |||
NR_073085.1 | 7 - 8 | 876 | 115 | |||
GenBank mRNA | AK057837.1 | - | 6 - 7 | 428 | 115 | |
AK091845.1 | - | 7 - 8 | 1404 | 115 | ||
AK223251.1 | - | 7 - 8 | 563 | 115 | ||
AK300092.1 | - | 7 - 8 | 1011 | 115 | ||
AK315757.1 | - | 7 - 8 | 603 | 115 | ||
BC000434.2 | - | 7 - 8 | 582 | 2819982 | 115 | |
BC001931.1 | - | 7 - 8 | 582 | 2819982 | 115 | |
BC034151.1 | - | 7 - 8 | 632 | 4690443 | 115 | |
BC128101.1 | - | 7 - 8 | 606 | 40109449 | 115 | |
BQ684851.1 | - | 241 | 115 | |||
BT019911.1 | - | 7 - 8 | 563 | 115 | ||
CN367966.1 | - | 6 - 7 | 514 | 115 | ||
D42123.1 | - | 7 - 8 | 589 | 115 | ||
U36190.1 | - | 7 - 8 | 599 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |