Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000624.5 | NP_000615.3 | 4 - 5 | 1125 | 71 | |
GenBank mRNA | AK096131.1 | - | 4 - 5 | 830 | 71 | |
AK293192.1 | - | 5 - 6 | 820 | 71 | ||
AK298279.1 | - | 4 - 5 | 731 | 71 | ||
AL080185.1 | - | 2 - 3 | 809 | 71 | ||
BC008915.2 | - | 4 - 5 | 1020 | 3618167 | 71 | |
J02639.1 | - | 3 - 4 | 936 | 71 | ||
S58545.1 | - | 2 - 3 | 812 | 71 | ||
U35464.1 | - | 2 - 3 | 914 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |