Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167681.2 | NP_001161153.1 | 7 - 8 | 1117 | 84 | |
NM_001271010.1 | NP_001257939.1 | 7 - 8 | 1234 | 84 | ||
NM_001271012.1 | NP_001257941.1 | 4 - 5 | 510 | 84 | ||
NM_002017.4 | NP_002008.2 | 6 - 7 | 1063 | 84 | ||
XM_011542701.2 | XP_011541003.1 | 6 - 7 | 878 | 84 | ||
XM_017017405.1 | XP_016872894.1 | 6 - 7 | 812 | 84 | ||
XM_017017406.1 | XP_016872895.1 | 6 - 7 | 1001 | 84 | ||
GenBank mRNA | AK300153.1 | - | 7 - 8 | 941 | 84 | |
AK313370.1 | - | 6 - 7 | 917 | 84 | ||
AY029368.1 | - | 6 - 7 | 720 | 84 | ||
BC001670.1 | - | 6 - 7 | 843 | 2989507 | 84 | |
BC010115.1 | - | 6 - 7 | 845 | 3635042 | 84 | |
M93255.1 | - | 7 - 8 | 1064 | 84 | ||
M98833.3 | - | 6 - 7 | 892 | 84 | ||
S45205.1 | - | 6 - 7 | 853 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |