Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000330701.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000330701.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000330701.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000330701.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000330701.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000330701.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000517702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000517702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000520584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000520584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000520584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000520584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000520584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000520584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000520584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000520584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000522233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
ENST00000522233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
NM_001271156.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
NM_001271156.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
NM_001271156.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
NM_001271156.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
NM_001271156.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
NM_001271156.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
NM_001271156.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
NM_001271156.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
NM_173832.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
NM_173832.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
NM_173832.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
NM_173832.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
NM_173832.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012373.hg.1 |
NM_173832.5 |
Refseq |