Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microtubule associated protein 10 | MAP10 |
KIAA1383, MTR120 |
-33.1kb | Sense | Chr. 1 - 232804892 - 232810346 on NCBI Build GRCh38 |
Downstream |
nucleoside-triphosphatase, cancer-related |
NTPCR |
C1orf57, HCR-NTPase |
92.1kb | Sense | Chr. 1 - 232950624 - 232978473 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |