Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 4287 | MIR4287 | -18.4kb | Anti-sense | Chr. 8 - 27886039 - 27886116 on NCBI Build GRCh38 | |
Downstream |
nuclear GTPase, germinal center associated |
NUGGC |
C8orf80, HMFN0672, SLIP-GC |
112.9kb | Anti-sense | Chr. 8 - 28021964 - 28084068 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |