Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000517428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000517428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000517428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000517428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000519893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000519893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000519893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000520256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000520256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000520256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
ENST00000520256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009900.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167239.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167239.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167239.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167239.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167239.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167239.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167239.hg.1 |
TC0800009900.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800167238.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167238.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167238.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167238.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167238.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167238.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167238.hg.1 |
TC0800009900.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000517428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000517428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000517428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000517428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000519364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000519893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000519893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000519893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000520256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000520256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000520256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
ENST00000520256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
NR_125809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009900.hg.1 |
NR_125809.1 |
Refseq |