Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300889.1 | NP_001287818.1 | 1 - 2 | 164 | 86 | |
GenBank mRNA | AY253917.1 | - | 1 - 2 | 122 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |