Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPCDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301101.1 | NP_001288030.1 | 4 - 5 | 548 | 105 | |
NM_001301102.1 | NP_001288031.1 | 4 - 5 | 581 | 105 | ||
NM_001301103.1 | NP_001288032.1 | 3 - 4 | 452 | 105 | ||
NM_001301104.1 | NP_001288033.1 | 3 - 4 | 570 | 105 | ||
NM_001301105.1 | NP_001288034.1 | 4 - 5 | 474 | 105 | ||
NM_021823.4 | NP_068595.3 | 5 - 6 | 677 | 105 | ||
NR_125369.1 | 4 - 5 | 470 | 105 | |||
GenBank mRNA | AF182419.1 | - | 3 - 4 | 553 | 105 | |
AK027491.1 | - | 5 - 6 | 639 | 105 | ||
AY358848.1 | - | 5 - 6 | 633 | 105 | ||
BC014409.1 | - | 4 - 5 | 603 | 4647763 | 105 | |
BI907452.1 | - | 4 - 5 | 401 | 105 | ||
BU500301.1 | - | 3 - 4 | 508 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |