Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029996.hg.1 |
TC01001934.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029996.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029996.hg.1 |
ENST00000366613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029996.hg.1 |
ENST00000366615.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029996.hg.1 |
ENST00000424237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029996.hg.1 |
NM_001012985.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029996.hg.1 |
NM_001206641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188648.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188648.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188648.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188648.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188648.hg.1 |
TC0100012020.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100188651.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188651.hg.1 |
ENST00000366613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188651.hg.1 |
ENST00000366615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188651.hg.1 |
ENST00000619305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188651.hg.1 |
NM_001012985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188651.hg.1 |
NM_001206641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188651.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188651.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188651.hg.1 |
ENST00000366613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188651.hg.1 |
ENST00000366615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188651.hg.1 |
ENST00000619305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188651.hg.1 |
NM_001012985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188651.hg.1 |
NM_001206641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188651.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188651.hg.1 |
TC0100012020.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000366615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
ENST00000619305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001012985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001206641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012020.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188652.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188652.hg.1 |
ENST00000366613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188652.hg.1 |
ENST00000366615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188652.hg.1 |
ENST00000619305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188652.hg.1 |
NM_001012985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188652.hg.1 |
NM_001206641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188652.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188652.hg.1 |
ENST00000366612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188652.hg.1 |
ENST00000366613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188652.hg.1 |
ENST00000366615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188652.hg.1 |
ENST00000619305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188652.hg.1 |
NM_001012985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188652.hg.1 |
NM_001206641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188652.hg.1 |
NM_001301733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188652.hg.1 |
TC0100012020.hg.1 |
Clariom D TC |