Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306149.1 | NP_001293078.1 | 2 - 3 | 466 | 82 | |
NM_001306150.1 | NP_001293079.1 | 2 - 3 | 466 | 82 | ||
NM_178569.3 | NP_848664.1 | 2 - 3 | 466 | 82 | ||
NR_122125.1 | 2 - 3 | 408 | 82 | |||
NR_122126.1 | 2 - 3 | 408 | 82 | |||
GenBank mRNA | AY249325.1 | - | 2 - 3 | 449 | 82 | |
BC101608.1 | - | 2 - 3 | 369 | 8069114 | 82 | |
BC101634.1 | - | 2 - 3 | 369 | 8069140 | 82 | |
BC143638.1 | - | 2 - 3 | 311 | 82 | ||
BC143639.1 | - | 2 - 3 | 369 | 9052153 | 82 | |
BC143640.1 | - | 2 - 3 | 311 | 82 | ||
CX754076.1 | - | 2 - 3 | 351 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |