Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010174.hg.1 |
ENST00000401675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010174.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010174.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010174.hg.1 |
NM_213720.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010174.hg.1 |
TC22000568.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010173.hg.1 |
ENST00000401675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010173.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010173.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010173.hg.1 |
NM_213720.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010173.hg.1 |
TC22000568.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010180.hg.1 |
ENST00000401675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010180.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010180.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010180.hg.1 |
NM_213720.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010180.hg.1 |
TC22000568.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010181.hg.1 |
ENST00000401675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010181.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010181.hg.1 |
NM_213720.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010181.hg.1 |
TC22000568.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010179.hg.1 |
ENST00000401675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010179.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010179.hg.1 |
NM_213720.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010179.hg.1 |
TC22000568.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000523865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000523865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000523865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
ENST00000523865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008186.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010177.hg.1 |
ENST00000401675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010177.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010177.hg.1 |
NM_213720.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010177.hg.1 |
TC22000568.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200158650.hg.1 |
TC2200008186.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200158650.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158650.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158650.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158650.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200158650.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158650.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158650.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158650.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200158648.hg.1 |
TC2200008186.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200158648.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158648.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158648.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158648.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158648.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200158648.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158648.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158648.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158648.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158648.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200158647.hg.1 |
TC2200008186.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200158647.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158647.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158647.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158647.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158643.hg.1 |
TC2200008186.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200158643.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158643.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158643.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158643.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158643.hg.1 |
ENST00000523865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158643.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200158643.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158643.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158643.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158643.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158643.hg.1 |
ENST00000523865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158643.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200158646.hg.1 |
TC2200008186.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200158646.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158646.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158646.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158646.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158646.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158646.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000401675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000484558.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000517886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000520222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000523865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000523865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000523865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
ENST00000523865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
NR_125756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008186.hg.1 |
NR_125756.1 |
Refseq |