Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000307358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000307358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000307358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000307358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000307358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000307358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000307358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000307358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000307358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000307358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000307358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000382512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000382512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000382512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000382512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000382512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000382512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000382512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
ENST00000382512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_001297559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_001297559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_001297559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_001297559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_001297559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_001297559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_001297559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_001297559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_001297559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_053044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_053044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_053044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_053044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_053044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_053044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_053044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_053044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_053044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_053044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006745.hg.1 |
NM_053044.4 |
Refseq |