Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225772.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225772.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225772.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225772.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225772.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225772.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225772.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225772.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225772.hg.1 |
TC0100016556.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
ENST00000367616.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_001297575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016556.hg.1 |
NM_014625.3 |
Refseq |