Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042549.1 | NP_001036014.1 | 2 - 3 | 346 | 114 | |
NM_001297736.1 | NP_001284665.1 | 2 - 3 | 346 | 114 | ||
NM_001297737.1 | NP_001284666.1 | 2 - 3 | 346 | 114 | ||
NM_001297739.1 | NP_001284668.1 | 2 - 3 | 346 | 114 | ||
NM_015471.3 | NP_056286.3 | 2 - 3 | 346 | 114 | ||
XM_017000968.1 | XP_016856457.1 | 3 - 4 | 1842 | 114 | ||
XM_017000969.1 | XP_016856458.1 | 3 - 4 | 1968 | 114 | ||
GenBank mRNA | AF201941.1 | - | 2 - 3 | 318 | 114 | |
AF255793.1 | - | 2 - 3 | 332 | 114 | ||
AI703127.1 | - | 2 - 3 | 130 | 114 | ||
AK222996.1 | - | 2 - 3 | 316 | 114 | ||
AK223260.1 | - | 2 - 3 | 326 | 114 | ||
AK313730.1 | - | 2 - 3 | 342 | 114 | ||
AL050084.1 | - | 2 - 3 | 125 | 114 | ||
BC007067.1 | - | 2 - 3 | 320 | 3997280 | 114 | |
BC036048.1 | - | 2 - 3 | 324 | 114 | ||
BE669670.1 | - | 2 - 3 | 151 | 114 | ||
BU570983.1 | - | 2 - 3 | 326 | 114 | ||
CK003923.1 | - | 2 - 3 | 329 | 114 | ||
DB045932.1 | - | 2 - 3 | 346 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |