Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000411527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000411527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000411527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000411527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000411527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000427449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000427449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000427449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000427449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000437316.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000437316.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000437316.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000437316.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000437316.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000437316.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011503.hg.1 |
ENST00000437316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000411527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000411527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000411527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000411527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000411527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000427449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000427449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000427449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000427449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000435145.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000437316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000437316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000437316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000437316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000437316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000437316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011503.hg.1 |
ENST00000437316.6 |
Ensembl |