Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000265284.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000376537.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000376544.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000376552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000428713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000462803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000463431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000467142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000470872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000490347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000495170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000496114.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
NM_001282748.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
NM_001282749.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
NM_001282753.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
NM_001282760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
NM_007005.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
NR_104239.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000265284.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000376537.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000376544.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000376552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000428713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000462803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000463431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000467142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000470872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000490347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000495170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
ENST00000496114.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
NM_001282748.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
NM_001282749.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
NM_001282753.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
NM_001282760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
NM_007005.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
NR_104239.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151956.hg.1 |
TC0900007680.hg.1 |
Clariom D TC |