Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
ENST00000274747.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
ENST00000378353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
ENST00000378386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
ENST00000443868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
ENST00000483634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
NM_001286264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
NM_001286265.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
NM_001286266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
NM_020662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
NR_104423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
ENST00000274747.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
ENST00000378353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
ENST00000378386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
ENST00000443868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
ENST00000483634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
NM_001286264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
NM_001286265.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
NM_001286266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
NM_020662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
NR_104423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148918.hg.1 |
TC0600007196.hg.1 |
Clariom D TC |