Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR7161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_106981.1 | 1 | 46 | 62 |
Gene Symbol: TMEM181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011536003.2 | XP_011534305.1 | 11 | 1767 | 62 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tubby like protein 4 | TULP4 | TUSP | -97.9kb | Sense | Chr. 6 - 158232192 - 158511828 on NCBI Build GRCh38 |
Downstream | dynein light chain Tctex-type 1 | DYNLT1 |
CW-1, TCTEL1, tctex-1 |
26.7kb | Anti-sense | Chr. 6 - 158636474 - 158644786 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |