Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000462431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000462431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000462431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000462431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000462431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000486168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000486168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000486168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000486168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196818.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196818.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196818.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196818.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196818.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196818.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196818.hg.1 |
TC0300012966.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000462431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000462431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000462431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000462431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000462431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000486168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000486168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000486168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000486168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
ENST00000497452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012966.hg.1 |
NR_108088.1 |
Refseq |