Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
ENST00000446290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
ENST00000446290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
ENST00000446290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
ENST00000446290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
NR_109769.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
NR_109769.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
NR_109769.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010323.hg.1 |
NR_109769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
ENST00000446290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
ENST00000446290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
ENST00000446290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
ENST00000446290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
NR_109769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
NR_109769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
NR_109769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010323.hg.1 |
NR_109769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900170718.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900170718.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900170718.hg.1 |
ENST00000446290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900170718.hg.1 |
NR_109769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900170718.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900170718.hg.1 |
ENST00000432148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900170718.hg.1 |
ENST00000446290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900170718.hg.1 |
NR_109769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900170718.hg.1 |
TC0900010323.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900170720.hg.1 |
ENST00000446290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900170720.hg.1 |
NR_109769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900170720.hg.1 |
ENST00000446290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900170720.hg.1 |
NR_109769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900170720.hg.1 |
TC0900010323.hg.1 |
Clariom D TC |