Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007543.hg.1 |
ENST00000506902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007543.hg.1 |
ENST00000506902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007543.hg.1 |
ENST00000506902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007543.hg.1 |
ENST00000506902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007543.hg.1 |
ENST00000506902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007543.hg.1 |
ENST00000506902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007543.hg.1 |
NR_109908.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007543.hg.1 |
NR_109908.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007543.hg.1 |
NR_109908.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007543.hg.1 |
NR_109908.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007543.hg.1 |
NR_109908.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007543.hg.1 |
NR_109908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007543.hg.1 |
ENST00000506902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007543.hg.1 |
ENST00000506902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007543.hg.1 |
ENST00000506902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007543.hg.1 |
ENST00000506902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007543.hg.1 |
ENST00000506902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007543.hg.1 |
ENST00000506902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007543.hg.1 |
NR_109908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007543.hg.1 |
NR_109908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007543.hg.1 |
NR_109908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007543.hg.1 |
NR_109908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007543.hg.1 |
NR_109908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007543.hg.1 |
NR_109908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500150832.hg.1 |
ENST00000506902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500150832.hg.1 |
NR_109908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500150832.hg.1 |
ENST00000506902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500150832.hg.1 |
NR_109908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500150832.hg.1 |
TC0500007543.hg.1 |
Clariom D TC |