Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000433969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000433969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000433969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000433969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000442719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000442719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000442719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000442719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000442719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000442719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_002331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_002331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_002331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_002331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_002331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_002331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_002331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_109980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_109980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_109980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_109980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_109980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_109981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_109981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_109981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_109981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_109981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012363.hg.1 |
NR_109981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189063.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189063.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189063.hg.1 |
NR_109980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189063.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189063.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189063.hg.1 |
NR_109980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189063.hg.1 |
TC0700012363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700189062.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189062.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189062.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189062.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189062.hg.1 |
NR_109980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189062.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189062.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189062.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189062.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189062.hg.1 |
NR_109980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189062.hg.1 |
TC0700012363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000432541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000433969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000433969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000433969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000433969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000434993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000442719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000442719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000442719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000442719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000442719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000442719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000446784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
ENST00000456577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_002331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_002331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_002331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_002331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_002331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_002331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_002331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_109980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_109980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_109980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_109980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_109980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_109981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_109981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_109981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_109981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_109981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012363.hg.1 |
NR_109981.1 |
Refseq |