Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110253.1 | 3 - 4 | 1212 | 104 | ||
XM_017007729.1 | XP_016863218.1 | 4 - 5 | 1278 | 104 | ||
XM_017007730.1 | XP_016863219.1 | 4 - 5 | 667 | 104 | ||
GenBank mRNA | BC109277.2 | - | 3 - 4 | 1088 | 104 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | THAP domain containing 9 | THAP9 | hTh9 | -19.6kb | Anti-sense | Chr. 4 - 82900650 - 82920283 on NCBI Build GRCh38 |
Downstream | COP9 signalosome subunit 4 | COPS4 | CSN4 | 64.6kb | Anti-sense | Chr. 4 - 83035086 - 83075818 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |