Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHKB-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110536.1 | 1 - 2 | 299 | 83 | ||
GenBank mRNA | BI561134.1 | - | 1 - 2 | 206 | 83 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | CHKB-CPT1B readthrough (NMD candidate) | CHKB-CPT1B | CHKL-CPT1B | -27.0 | Anti-sense | Chr. 22 - 50568861 - 50582999 on NCBI Build GRCh38 |
Downstream | mitogen-activated protein kinase 8 interacting protein 2 | MAPK8IP2 |
IB-2, IB2, JIP2, PRKM8IPL |
13.7kb | Sense | Chr. 22 - 50600685 - 50613978 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |