Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015725.hg.1 |
ENST00000425481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015725.hg.1 |
ENST00000425481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015725.hg.1 |
ENST00000425481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015725.hg.1 |
ENST00000441749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015725.hg.1 |
ENST00000441749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015725.hg.1 |
ENST00000441749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015725.hg.1 |
NR_110573.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015725.hg.1 |
NR_110573.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015725.hg.1 |
NR_110573.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015725.hg.1 |
NR_110573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200215436.hg.1 |
ENST00000425481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215436.hg.1 |
ENST00000441749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215436.hg.1 |
NR_110573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200215436.hg.1 |
ENST00000425481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215436.hg.1 |
ENST00000441749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215436.hg.1 |
NR_110573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200215436.hg.1 |
TC0200015725.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200215438.hg.1 |
ENST00000425481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215438.hg.1 |
ENST00000441749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215438.hg.1 |
NR_110573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200215438.hg.1 |
ENST00000425481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215438.hg.1 |
ENST00000441749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215438.hg.1 |
NR_110573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200215438.hg.1 |
TC0200015725.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200215442.hg.1 |
NR_110573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200215442.hg.1 |
NR_110573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200215442.hg.1 |
TC0200015725.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200215441.hg.1 |
ENST00000441749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215441.hg.1 |
NR_110573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200215441.hg.1 |
ENST00000441749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215441.hg.1 |
NR_110573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200215441.hg.1 |
TC0200015725.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015725.hg.1 |
ENST00000425481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015725.hg.1 |
ENST00000425481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015725.hg.1 |
ENST00000425481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015725.hg.1 |
ENST00000441749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015725.hg.1 |
ENST00000441749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015725.hg.1 |
ENST00000441749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015725.hg.1 |
NR_110573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015725.hg.1 |
NR_110573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015725.hg.1 |
NR_110573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015725.hg.1 |
NR_110573.1 |
Refseq |