Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
ENST00000433008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
ENST00000433008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
ENST00000433008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
NR_117098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
NR_117098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
NR_117098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
NR_117098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
NR_117098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011252.hg.1 |
NR_117098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182620.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182620.hg.1 |
ENST00000433008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182620.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182620.hg.1 |
NR_117098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182620.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182620.hg.1 |
ENST00000433008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182620.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182620.hg.1 |
NR_117098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182620.hg.1 |
TC0100011252.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
ENST00000433008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
ENST00000433008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
ENST00000433008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
NR_117098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
NR_117098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
NR_117098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
NR_117098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
NR_117098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011252.hg.1 |
NR_117098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182617.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182617.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182617.hg.1 |
ENST00000412772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182617.hg.1 |
NR_117097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182617.hg.1 |
TC0100011252.hg.1 |
Clariom D TC |