Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
ENST00000310999.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
ENST00000329819.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
ENST00000430466.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
ENST00000524576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
ENST00000534488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
NM_016050.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
NM_170738.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
NM_170739.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
NR_120588.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
NR_120589.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
ENST00000310999.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
ENST00000329819.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
ENST00000430466.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
ENST00000524576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
ENST00000534488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
NM_016050.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
NM_170738.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
NM_170739.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
NR_120588.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
NR_120589.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186878.hg.1 |
TC1100011288.hg.1 |
Clariom D TC |